Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CHRFAM7A
- Entrez Gene ID
-
89832
- Gene Name
- CHRNA7 (exons 5-10) and FAM7A (exons A-E) fusion
- Gene Aliases
-
CHRNA7, CHRNA7-DR1, D-10
- Chromosome Location
-
Chr.15: 30357770 - 30393849 on Build GRCh38
- Gene Symbol
- ULK4P3
- Entrez Gene ID
-
89837
- Gene Name
- ULK4 pseudogene 3
- Gene Aliases
-
D-X, FAM7A3
- Chromosome Location
-
Chr.15: 30103732 - 30131748 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_026859.1 |
|
1 - 2 |
270 |
88 |
GenBank mRNA |
BC023564.2 |
- |
1 - 2 |
270 |
88 |
BC071855.1 |
- |
1 - 2 |
268 |
88 |
View Full Product Details