The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013579.2 | NP_001013597.1 | 4 - 5 | 506 | 95 |
XM_017029295.1 | XP_016884784.1 | 4 - 5 | 1672 | 95 | |
GenBank mRNA | AY947638.1 | - | 4 - 5 | 465 | 95 |