The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161528.1 | NP_001155000.1 | 2 - 3 | 2191 | 124 |
GenBank mRNA | BC026112.1 | - | 2 - 3 | 727 | 124 |
BC144674.1 | - | 2 - 3 | 286 | 124 | |
BC144678.1 | - | 2 - 3 | 309 | 124 |