Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLSCR2
Entrez Gene ID
57047
Gene Name
phospholipid scramblase 2
Gene Aliases
-
Chromosome Location
Chr.3: 146391363 - 146497163 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011513013.2 XP_011511315.2 11 - 12 5190 121
XM_011513019.2 XP_011511321.2 10 - 11 2550 121
XM_011513020.2 XP_011511322.2 11 - 12 2613 121
XM_011513021.2 XP_011511323.2 9 - 10 2350 121
XM_017006898.1 XP_016862387.1 12 - 13 3429 121
XM_017006899.1 XP_016862388.1 12 - 13 5253 121
XM_017006900.1 XP_016862389.1 11 - 12 3326 121
XM_017006901.1 XP_016862390.1 12 - 13 5063 121
XM_017006902.1 XP_016862391.1 12 - 13 3389 121
XM_017006905.1 XP_016862394.1 13 - 14 4646 121
XM_017006906.1 XP_016862395.1 13 - 14 4427 121
XM_017006907.1 XP_016862396.1 13 - 14 4192 121
XM_017006908.1 XP_016862397.1 13 - 14 4317 121
XM_017006909.1 XP_016862398.1 12 - 13 4104 121
XM_017006912.1 XP_016862401.1 11 - 12 3845 121
XM_017006914.1 XP_016862403.1 11 - 12 4052 121
XM_017006915.1 XP_016862404.1 11 - 12 4034 121

View Full Product Details