Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL28A1
Entrez Gene ID
340267
Gene Name
collagen type XXVIII alpha 1 chain
Gene Aliases
COL28
Chromosome Location
Chr.7: 7338543 - 7542062 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037763.2 NP_001032852.2 33 - 34 3185 85
XM_011515358.2 XP_011513660.1 33 - 34 8030 85
XM_011515359.2 XP_011513661.1 33 - 34 3188 85
XM_011515360.2 XP_011513662.1 33 - 34 4470 85
XM_011515362.2 XP_011513664.1 21 - 22 1990 85
XM_017012131.1 XP_016867620.1 34 - 35 9206 85
XM_017012132.1 XP_016867621.1 33 - 34 4470 85
GenBank mRNA AJ890451.1 - 33 - 34 3185 85
BC063866.1 - 1 - 2 119 85
BC136892.1 - 33 - 34 3175 85
BC144431.1 - 34 - 35 3360 85

View Full Product Details