Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- BANP
- Entrez Gene ID
-
54971
- Gene Name
- BTG3 associated nuclear protein
- Gene Aliases
-
BEND1, SMAR1, SMARBP1
- Chromosome Location
-
Chr.16: 87949216 - 88077318 on Build GRCh38
View Full Product Details