Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 243316632-243317187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDCCAG8
Entrez Gene ID
10806
Gene Name
serologically defined colon cancer antigen 8
Gene Aliases
BBS16, CCCAP, CCCAP SLSN7, HSPC085, NPHP10, NY-CO-8, SLSN7, hCCCAP
UniGene
Hs.591530
Gene Chromosome Location
Chr.1: 243255328-243500092 on Build GRCh38
Transcripts
NM_006642.3 XM_005273013.4 XM_005273018.1 XM_005273021.4 XM_005273022.3 XM_005273023.4 XM_006711727.3 XM_006711728.3 XM_011544021.2 XM_011544022.2 XM_011544023.2 XM_011544024.2 XM_011544026.2 XM_011544028.2 XM_011544029.2 XM_011544030.2 XM_017000103.1 XM_017000104.1 XM_017000105.1 XM_017000106.1 XM_017000107.1 XM_017000108.1 XM_017000109.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4519640 c.1020G>A p.V340V Chr.1: 243316845-243316845 on Build GRCh38
5053294 c.1035A>G p.Q345Q Chr.1: 243316860-243316860 on Build GRCh38
1296093 c.953C>T p.A318V Chr.1: 243316778-243316778 on Build GRCh38
4030240 c.934A>G p.R312G Chr.1: 243316759-243316759 on Build GRCh38
906601 c.1027G>T p.V343F Chr.1: 243316852-243316852 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774019484 Chr.1: 243316913-243316915 on Build GRCh38
rs752228630 Chr.1: 243316920-243316920 on Build GRCh38
rs780738331 Chr.1: 243316795-243316795 on Build GRCh38
rs770394700 Chr.1: 243316820-243316820 on Build GRCh38
rs773312170 Chr.1: 243316800-243316800 on Build GRCh38
rs143807801 Chr.1: 243316712-243316712 on Build GRCh38
rs951295161 Chr.1: 243316853-243316853 on Build GRCh38
rs547714378 Chr.1: 243316732-243316732 on Build GRCh38
rs111775515 Chr.1: 243316766-243316766 on Build GRCh38
rs995846717 Chr.1: 243316697-243316697 on Build GRCh38
rs199524638 Chr.1: 243316889-243316889 on Build GRCh38
rs562212927 Chr.1: 243317088-243317090 on Build GRCh38
rs559121131 Chr.1: 243316928-243316929 on Build GRCh38
rs527870268 Chr.1: 243316875-243316875 on Build GRCh38
rs753594667 Chr.1: 243316934-243316934 on Build GRCh38
rs746449124 Chr.1: 243316822-243316822 on Build GRCh38
rs6672843 Chr.1: 243316789-243316789 on Build GRCh38
rs909766418 Chr.1: 243316669-243316669 on Build GRCh38
rs906876743 Chr.1: 243317163-243317163 on Build GRCh38
rs137903012 Chr.1: 243316734-243316734 on Build GRCh38
rs984454237 Chr.1: 243316668-243316668 on Build GRCh38
rs200294385 Chr.1: 243316769-243316769 on Build GRCh38
rs755672794 Chr.1: 243316925-243316925 on Build GRCh38
rs892211642 Chr.1: 243317028-243317028 on Build GRCh38
rs754561913 Chr.1: 243316708-243316708 on Build GRCh38
rs372404255 Chr.1: 243316780-243316780 on Build GRCh38
rs780791623 Chr.1: 243316709-243316709 on Build GRCh38
rs1039938968 Chr.1: 243316948-243316948 on Build GRCh38
rs899970728 Chr.1: 243316956-243316956 on Build GRCh38
rs777038486 Chr.1: 243316878-243316878 on Build GRCh38
rs563410283 Chr.1: 243317141-243317141 on Build GRCh38
rs762472937 Chr.1: 243316811-243316812 on Build GRCh38
rs564475323 Chr.1: 243316910-243316910 on Build GRCh38
rs753005735 Chr.1: 243316929-243316929 on Build GRCh38
rs771991355 Chr.1: 243316835-243316835 on Build GRCh38
rs751551796 Chr.1: 243316788-243316788 on Build GRCh38
rs767858763 Chr.1: 243316925-243316935 on Build GRCh38
rs1004676048 Chr.1: 243317127-243317127 on Build GRCh38
rs77555777 Chr.1: 243316921-243316921 on Build GRCh38
rs777374457 Chr.1: 243316926-243316926 on Build GRCh38
rs767707839 Chr.1: 243316759-243316759 on Build GRCh38
rs759174810 Chr.1: 243316916-243316917 on Build GRCh38
rs909838385 Chr.1: 243316924-243316924 on Build GRCh38
rs576672580 Chr.1: 243316793-243316793 on Build GRCh38
rs980428411 Chr.1: 243316947-243316947 on Build GRCh38
rs201388041 Chr.1: 243316896-243316896 on Build GRCh38
rs944285202 Chr.1: 243316945-243316945 on Build GRCh38
rs35859404 Chr.1: 243316811-243316811 on Build GRCh38
rs142658750 Chr.1: 243316821-243316821 on Build GRCh38
rs927652402 Chr.1: 243316950-243316953 on Build GRCh38
rs887509741 Chr.1: 243317125-243317125 on Build GRCh38
rs747505682 Chr.1: 243316800-243316800 on Build GRCh38
rs750272621 Chr.1: 243316779-243316779 on Build GRCh38
rs929799852 Chr.1: 243316666-243316666 on Build GRCh38
rs778065772 Chr.1: 243316812-243316812 on Build GRCh38
rs145984257 Chr.1: 243316847-243316847 on Build GRCh38
rs889392770 Chr.1: 243317120-243317120 on Build GRCh38
rs1043885705 Chr.1: 243317088-243317088 on Build GRCh38
rs759504214 Chr.1: 243316753-243316753 on Build GRCh38
rs931498158 Chr.1: 243316959-243316959 on Build GRCh38
rs760942783 Chr.1: 243316852-243316852 on Build GRCh38
rs200461035 Chr.1: 243316772-243316772 on Build GRCh38
rs759530315 Chr.1: 243316911-243316911 on Build GRCh38
rs912601625 Chr.1: 243316941-243316941 on Build GRCh38
rs543631756 Chr.1: 243317017-243317017 on Build GRCh38
rs749594870 Chr.1: 243316817-243316817 on Build GRCh38
rs747862454 Chr.1: 243316710-243316710 on Build GRCh38
rs1014996850 Chr.1: 243317138-243317138 on Build GRCh38
rs1014619954 Chr.1: 243316738-243316738 on Build GRCh38
rs900042782 Chr.1: 243316695-243316695 on Build GRCh38
rs1048775652 Chr.1: 243317006-243317006 on Build GRCh38
rs371778586 Chr.1: 243316747-243316747 on Build GRCh38
rs771368318 Chr.1: 243316818-243316818 on Build GRCh38
rs766944780 Chr.1: 243316916-243316916 on Build GRCh38
rs535395368 Chr.1: 243316828-243316828 on Build GRCh38
rs769785828 Chr.1: 243316712-243316712 on Build GRCh38
rs977804626 Chr.1: 243316937-243316937 on Build GRCh38
rs368258515 Chr.1: 243316775-243316775 on Build GRCh38
rs577794803 Chr.1: 243317001-243317001 on Build GRCh38
rs774972770 Chr.1: 243316751-243316751 on Build GRCh38

View Full Product Details