Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHCHD3
Entrez Gene ID
54927
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 3
Gene Aliases
MINOS3, Mic19, PPP1R22
Chromosome Location
Chr.7: 132784862 - 133082158 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317177.1 NP_001304106.1 8 - 9 898 86
NM_017812.3 NP_060282.1 7 - 8 883 86
NR_133671.1 10 - 11 1264 86
GenBank mRNA AK000427.1 - 7 - 8 799 86
AK129533.1 - 7 - 8 764 86
AK310236.1 - 10 - 11 1176 86
BC011596.2 - 7 - 8 805 86
BC014839.1 - 7 - 8 807 86

View Full Product Details