The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020770.2 | NP_065821.1 | 11 - 12 | 2247 | 54 |
XM_005245365.4 | XP_005245422.1 | 11 - 12 | 2259 | 54 | |
GenBank mRNA | AB037740.1 | - | 11 - 12 | 2220 | 54 |
AF263462.1 | - | 11 - 12 | 2254 | 54 | |
AK290007.1 | - | 11 - 12 | 2220 | 54 | |
BC150604.1 | - | 10 - 11 | 2126 | 54 | |
BC152445.1 | - | 11 - 12 | 2220 | 54 |