Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CGN
Entrez Gene ID
57530
Gene Name
cingulin
Gene Aliases
-
Chromosome Location
Chr.1: 151510859 - 151538692 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020770.2 NP_065821.1 2 - 3 1012 90
XM_005245365.4 XP_005245422.1 2 - 3 1024 90
GenBank mRNA AB037740.1 - 2 - 3 985 90
AF263462.1 - 2 - 3 1019 90
AK290007.1 - 2 - 3 985 90
BC146657.1 - 2 - 3 941 90
BC150604.1 - 1 - 2 891 90
BC152445.1 - 2 - 3 985 90

View Full Product Details