The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020770.2 | NP_065821.1 | 4 - 5 | 1185 | 62 |
XM_005245365.4 | XP_005245422.1 | 4 - 5 | 1197 | 62 | |
GenBank mRNA | AB037740.1 | - | 4 - 5 | 1158 | 62 |
AF263462.1 | - | 4 - 5 | 1192 | 62 | |
AK290007.1 | - | 4 - 5 | 1158 | 62 | |
BC146657.1 | - | 4 - 5 | 1114 | 62 | |
BC150604.1 | - | 3 - 4 | 1064 | 62 | |
BC152445.1 | - | 4 - 5 | 1158 | 62 |