Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 161349068-161349654 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PARK2
Entrez Gene ID
5071
Gene Name
parkin RBR E3 ubiquitin protein ligase
Gene Aliases
AR-JP, LPRS2, PDJ, PRKN
UniGene
Hs.132954
Gene Chromosome Location
Chr.6: 161347417-162727802 on Build GRCh38
Transcripts
NM_004562.2 NM_013987.2 NM_013988.2 XM_011535863.1 XM_017010908.1 XM_017010909.1

SNP Detail

SNP ID Chromosome Location
rs572345942 Chr.6: 161349209-161349212 on Build GRCh38
rs957788053 Chr.6: 161349154-161349154 on Build GRCh38
rs1032474559 Chr.6: 161349150-161349150 on Build GRCh38
rs1049142631 Chr.6: 161349194-161349194 on Build GRCh38
rs774941206 Chr.6: 161349258-161349258 on Build GRCh38
rs565359877 Chr.6: 161349470-161349470 on Build GRCh38
rs541658812 Chr.6: 161349220-161349220 on Build GRCh38
rs760233938 Chr.6: 161349446-161349446 on Build GRCh38
rs112757570 Chr.6: 161349571-161349571 on Build GRCh38
rs902436659 Chr.6: 161349456-161349456 on Build GRCh38
rs1047898450 Chr.6: 161349312-161349312 on Build GRCh38
rs528792599 Chr.6: 161349424-161349424 on Build GRCh38
rs965011358 Chr.6: 161349502-161349502 on Build GRCh38
rs1039492436 Chr.6: 161349121-161349121 on Build GRCh38
rs776047624 Chr.6: 161349369-161349369 on Build GRCh38
rs943928519 Chr.6: 161349622-161349622 on Build GRCh38
rs71653629 Chr.6: 161349447-161349447 on Build GRCh38
rs912250905 Chr.6: 161349590-161349590 on Build GRCh38
rs1014955697 Chr.6: 161349276-161349276 on Build GRCh38
rs942110218 Chr.6: 161349325-161349325 on Build GRCh38
rs988197174 Chr.6: 161349544-161349544 on Build GRCh38
rs1055099691 Chr.6: 161349352-161349352 on Build GRCh38
rs567299724 Chr.6: 161349515-161349515 on Build GRCh38
rs530999210 Chr.6: 161349476-161349476 on Build GRCh38
rs573709424 Chr.6: 161349196-161349196 on Build GRCh38
rs996187483 Chr.6: 161349188-161349188 on Build GRCh38
rs776532449 Chr.6: 161349558-161349561 on Build GRCh38
rs966945271 Chr.6: 161349111-161349111 on Build GRCh38
rs1030964785 Chr.6: 161349542-161349542 on Build GRCh38
rs767963623 Chr.6: 161349287-161349287 on Build GRCh38
rs909255211 Chr.6: 161349318-161349318 on Build GRCh38
rs1032155646 Chr.6: 161349469-161349469 on Build GRCh38
rs74701717 Chr.6: 161349279-161349279 on Build GRCh38
rs969554917 Chr.6: 161349404-161349404 on Build GRCh38
rs1006582328 Chr.6: 161349484-161349484 on Build GRCh38
rs899692434 Chr.6: 161349178-161349178 on Build GRCh38
rs530081303 Chr.6: 161349549-161349549 on Build GRCh38
rs753440085 Chr.6: 161349573-161349573 on Build GRCh38
rs1013865134 Chr.6: 161349284-161349284 on Build GRCh38
rs999729613 Chr.6: 161349466-161349466 on Build GRCh38
rs943996494 Chr.6: 161349115-161349115 on Build GRCh38
rs886061235 Chr.6: 161349627-161349627 on Build GRCh38
rs188421588 Chr.6: 161349491-161349491 on Build GRCh38
rs773701392 Chr.6: 161349208-161349209 on Build GRCh38
rs895132585 Chr.6: 161349376-161349376 on Build GRCh38
rs910901981 Chr.6: 161349202-161349202 on Build GRCh38

View Full Product Details