Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47216923-47217095 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDB2
Entrez Gene ID
1643
Gene Name
damage specific DNA binding protein 2
Gene Aliases
DDBB, UV-DDB2, XPE
UniGene
Hs.700338
Gene Chromosome Location
Chr.11: 47214942-47239218 on Build GRCh38
Transcripts
NM_000107.2 NM_001300734.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4033131 c.382G>A p.A128T Chr.11: 47216975-47216975 on Build GRCh38
6340893 c.433G>T p.D145Y Chr.11: 47217026-47217026 on Build GRCh38
1981294 c.379G>A p.V127M Chr.11: 47216972-47216972 on Build GRCh38
6387619 c.449T>G p.I150S Chr.11: 47217042-47217042 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762943470 Chr.11: 47216963-47216963 on Build GRCh38
rs549726695 Chr.11: 47217057-47217057 on Build GRCh38
rs145264146 Chr.11: 47217043-47217043 on Build GRCh38
rs769144378 Chr.11: 47216946-47216946 on Build GRCh38
rs201531216 Chr.11: 47217069-47217069 on Build GRCh38
rs769321481 Chr.11: 47216958-47216958 on Build GRCh38
rs747546269 Chr.11: 47217066-47217066 on Build GRCh38
rs750530703 Chr.11: 47217036-47217036 on Build GRCh38
rs766359606 Chr.11: 47216969-47216969 on Build GRCh38
rs776380322 Chr.11: 47216947-47216947 on Build GRCh38
rs139968905 Chr.11: 47216991-47216991 on Build GRCh38
rs772637982 Chr.11: 47216962-47216962 on Build GRCh38
rs767324814 Chr.11: 47216972-47216972 on Build GRCh38
rs747774067 Chr.11: 47216950-47216950 on Build GRCh38
rs200856208 Chr.11: 47216993-47216993 on Build GRCh38
rs757461854 Chr.11: 47217001-47217001 on Build GRCh38
rs765357628 Chr.11: 47217028-47217028 on Build GRCh38
rs326212 Chr.11: 47216971-47216971 on Build GRCh38

View Full Product Details