Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 66443204-66443331 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP2K1
Entrez Gene ID
5604
Gene Name
mitogen-activated protein kinase kinase 1
Gene Aliases
CFC3, MAPKK1, MEK1, MKK1, PRKMK1
UniGene
Hs.145442
Gene Chromosome Location
Chr.15: 66386873-66491544 on Build GRCh38
Transcripts
NM_002755.3 XM_011521783.2 XM_017022411.1 XM_017022412.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5894998 c.439-5C>T p.? Chr.15: 66443275-66443275 on Build GRCh38
3690651 c.449C>T p.S150F Chr.15: 66443290-66443290 on Build GRCh38
1724124 c.484C>T p.P162S Chr.15: 66443325-66443325 on Build GRCh38
6968948 c.473C>T p.A158V Chr.15: 66443314-66443314 on Build GRCh38
5849601 c.460G>A p.V154I Chr.15: 66443301-66443301 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758059146 Chr.15: 66443258-66443258 on Build GRCh38
rs747709090 Chr.15: 66443272-66443272 on Build GRCh38
rs751155864 Chr.15: 66443262-66443262 on Build GRCh38
rs886066297 Chr.15: 66443224-66443224 on Build GRCh38
rs754725417 Chr.15: 66443266-66443266 on Build GRCh38
rs766755726 Chr.15: 66443256-66443256 on Build GRCh38
rs1051316105 Chr.15: 66443245-66443245 on Build GRCh38
rs964442363 Chr.15: 66443273-66443273 on Build GRCh38
rs34839715 Chr.15: 66443231-66443232 on Build GRCh38
rs755398652 Chr.15: 66443294-66443294 on Build GRCh38
rs779497093 Chr.15: 66443282-66443282 on Build GRCh38
rs780816788 Chr.15: 66443271-66443271 on Build GRCh38
rs766207567 Chr.15: 66443261-66443261 on Build GRCh38
rs750386919 Chr.15: 66443246-66443246 on Build GRCh38
rs751939652 Chr.15: 66443260-66443261 on Build GRCh38
rs755707394 Chr.15: 66443280-66443280 on Build GRCh38

View Full Product Details