Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 55143368-55143537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGFR
Entrez Gene ID
1956
Gene Name
epidermal growth factor receptor
Gene Aliases
ERBB, ERBB1, HER1, NISBD2, PIG61, mENA
UniGene
Hs.488293
Gene Chromosome Location
Chr.7: 55019032-55207338 on Build GRCh38
Transcripts
NM_005228.3 NM_201282.1 NM_201283.1 NM_201284.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6939064 c.322A>G p.R108G Chr.7: 55143386-55143386 on Build GRCh38
6962471 c.412A>G p.R138G Chr.7: 55143476-55143476 on Build GRCh38
6191491 c.311T>A p.L104Q Chr.7: 55143375-55143375 on Build GRCh38
6978221 c.413G>T p.R138I Chr.7: 55143477-55143477 on Build GRCh38
5463644 c.405G>A p.L135L Chr.7: 55143469-55143469 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142553829 Chr.7: 55143436-55143436 on Build GRCh38
rs746631025 Chr.7: 55143396-55143396 on Build GRCh38
rs756250461 Chr.7: 55143506-55143506 on Build GRCh38
rs374582814 Chr.7: 55143403-55143403 on Build GRCh38
rs762889647 Chr.7: 55143450-55143450 on Build GRCh38
rs147726446 Chr.7: 55143418-55143418 on Build GRCh38
rs767608234 Chr.7: 55143487-55143487 on Build GRCh38
rs751337426 Chr.7: 55143464-55143464 on Build GRCh38
rs753033730 Chr.7: 55143491-55143491 on Build GRCh38
rs377444977 Chr.7: 55143443-55143443 on Build GRCh38
rs773596817 Chr.7: 55143409-55143409 on Build GRCh38
rs764128212 Chr.7: 55143491-55143492 on Build GRCh38
rs944455554 Chr.7: 55143509-55143509 on Build GRCh38
rs766093262 Chr.7: 55143454-55143454 on Build GRCh38
rs754854319 Chr.7: 55143473-55143473 on Build GRCh38

View Full Product Details