Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HCRT
Entrez Gene ID
3060
Gene Name
hypocretin neuropeptide precursor
Gene Aliases
NRCLP1, OX, PPOX
Chromosome Location
Chr.17: 42184060 - 42185452 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001524.1 NP_001515.1 1 - 2 109 101
GenBank mRNA AF041240.1 - 1 - 2 109 101

View Full Product Details