Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- IFNA14
- Entrez Gene ID
-
3448
- Gene Name
- interferon alpha 14
- Gene Aliases
-
IFN-alphaH, LEIF2H
- Chromosome Location
-
Chr.9: 21239202 - 21239979 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
V00542.1 |
- |
1 - 1 |
42 |
71 |
View Full Product Details