The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002812.4 | NP_002803.2 | 5 - 6 | 861 | 105 |
GenBank mRNA | AK301771.1 | - | 5 - 6 | 872 | 105 |
AK304486.1 | - | 5 - 6 | 446 | 105 | |
AK308809.1 | - | 5 - 6 | 950 | 105 | |
BC001164.1 | - | 5 - 6 | 612 | 105 | |
BC065006.1 | - | 5 - 6 | 600 | 105 | |
BQ684248.1 | - | 5 - 6 | 624 | 105 | |
BT019688.1 | - | 5 - 6 | 530 | 105 | |
D38047.1 | - | 5 - 6 | 600 | 105 | |
EU668354.1 | - | 5 - 6 | 861 | 105 |