The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032444.2 | NP_115820.2 | 12 - 13 | 5264 | 76 |
XM_017023775.1 | XP_016879264.1 | 9 - 10 | 4669 | 76 | |
GenBank mRNA | AB058687.1 | - | - | 2638 | 76 |
AK095411.1 | - | 1 - 2 | 1134 | 76 | |
BC036335.1 | - | 3 - 4 | 2656 | 76 |