Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNRNP200
Entrez Gene ID
23020
Gene Name
small nuclear ribonucleoprotein U5 subunit 200
Gene Aliases
ASCC3L1, BRR2, HELIC2, RP33, U5-200KD
Chromosome Location
Chr.2: 96274336 - 96321893 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014014.4 NP_054733.2 12 - 13 1647 58
XM_017003603.1 XP_016859092.1 14 - 15 2477 58
GenBank mRNA AB018331.2 - 10 - 11 1185 58
AK024391.1 - 6 - 7 728 58
AK090671.1 - 12 - 13 1624 58
AL831994.1 - 3 - 4 363 58
AY572488.1 - 12 - 13 1593 58
BC065924.1 - 5 - 6 570 58
BC112891.1 - 6 - 7 768 58

View Full Product Details