The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145364.2 | NP_001138836.1 | 2 - 3 | 364 | 103 |
NM_001301058.1 | NP_001287987.1 | 2 - 3 | 364 | 103 | |
NM_001321383.1 | NP_001308312.1 | 2 - 3 | 364 | 103 | |
NM_148912.3 | NP_683710.1 | 2 - 3 | 364 | 103 | |
NM_148913.3 | NP_683711.1 | 2 - 3 | 343 | 103 | |
NR_135627.1 | 2 - 3 | 364 | 103 | ||
GenBank mRNA | AF412030.1 | - | 2 - 3 | 356 | 103 |
AY053499.1 | - | 2 - 3 | 335 | 103 | |
AY053500.1 | - | 2 - 3 | 335 | 103 | |
BC008251.1 | - | 2 - 3 | 328 | 103 | |
BC011712.2 | - | 2 - 3 | 282 | 103 | |
BC067750.1 | - | 2 - 3 | 299 | 103 | |
BM557977.1 | - | 2 - 3 | 364 | 103 | |
BU175566.1 | - | 2 - 3 | 247 | 103 | |
BU189995.1 | - | 2 - 3 | 320 | 103 | |
CB998040.1 | - | 2 - 3 | 356 | 103 | |
DB460434.1 | - | 2 - 3 | 357 | 103 |