Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PROSER2
Entrez Gene ID
254427
Gene Name
proline and serine rich 2
Gene Aliases
C10orf47
Chromosome Location
Chr.10: 11823341 - 11872277 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153256.3 NP_694988.3 4 - 4 1454 102
XM_011519438.2 XP_011517740.1 5 - 5 1773 102
XM_017016028.1 XP_016871517.1 4 - 4 2440 102
GenBank mRNA BC007547.1 - 1 - 1 372 102
BC017269.2 - 5 - 5 1172 102
BC029134.1 - 1 - 1 755 102
BC048800.1 - 4 - 4 1454 102

View Full Product Details