Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CYP4Z1
- Entrez Gene ID
-
199974
- Gene Name
- cytochrome P450 family 4 subfamily Z member 1
- Gene Aliases
-
CYP4A20
- Chromosome Location
-
Chr.1: 47067488 - 47118320 on Build GRCh38
- Gene Symbol
- CYP4Z2P
- Entrez Gene ID
-
163720
- Gene Name
- cytochrome P450 family 4 subfamily Z member 2, pseudogene
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 46843095 - 46900475 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002788.2 |
|
5 - 6 |
654 |
82 |
GenBank mRNA |
AK097373.1 |
- |
5 - 6 |
654 |
82 |
AY262057.1 |
- |
5 - 6 |
616 |
82 |
AY696295.1 |
- |
5 - 6 |
613 |
82 |
View Full Product Details