The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282560.1 | NP_001269489.1 | 4 - 5 | 337 | 99 |
NM_001282561.1 | NP_001269490.1 | 4 - 5 | 337 | 99 | |
NM_001990.3 | NP_001981.2 | 4 - 5 | 337 | 99 | |
NR_104214.1 | 4 - 5 | 337 | 99 | ||
XM_006710449.3 | XP_006710512.1 | 5 - 6 | 373 | 99 | |
XM_011540999.2 | XP_011539301.1 | 5 - 6 | 376 | 99 | |
XM_011541001.2 | XP_011539303.1 | 4 - 5 | 334 | 99 | |
XM_011541003.2 | XP_011539305.1 | 5 - 6 | 374 | 99 | |
XM_011541004.2 | XP_011539306.1 | 5 - 6 | 371 | 99 | |
XM_017000652.1 | XP_016856141.1 | 5 - 6 | 377 | 99 | |
XM_017000653.1 | XP_016856142.1 | 6 - 7 | 497 | 99 | |
XM_017000654.1 | XP_016856143.1 | 4 - 5 | 334 | 99 | |
XM_017000655.1 | XP_016856144.1 | 5 - 6 | 490 | 99 | |
XM_017000656.1 | XP_016856145.1 | 5 - 6 | 375 | 99 | |
XM_017000657.1 | XP_016856146.1 | 4 - 5 | 367 | 99 | |
GenBank mRNA | AK289805.1 | - | 4 - 5 | 296 | 99 |
AK295745.1 | - | 4 - 5 | 331 | 99 | |
AL603098.1 | - | 4 - 5 | 320 | 99 | |
BC029500.1 | - | 4 - 5 | 297 | 99 | |
BC041667.1 | - | 4 - 5 | 264 | 99 | |
BX648945.1 | - | 4 - 5 | 306 | 99 | |
DC416048.1 | - | 4 - 5 | 337 | 99 |