Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EYA3
Entrez Gene ID
2140
Gene Name
EYA transcriptional coactivator and phosphatase 3
Gene Aliases
-
Chromosome Location
Chr.1: 27970344 - 28088671 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282560.1 NP_001269489.1 5 - 6 405 100
NM_001282561.1 NP_001269490.1 5 - 6 405 100
NM_001282562.1 NP_001269491.1 4 - 5 325 100
NM_001990.3 NP_001981.2 5 - 6 405 100
NR_104214.1 5 - 6 405 100
XM_006710449.3 XP_006710512.1 6 - 7 441 100
XM_011540999.2 XP_011539301.1 6 - 7 444 100
XM_011541001.2 XP_011539303.1 5 - 6 402 100
XM_011541003.2 XP_011539305.1 6 - 7 442 100
XM_011541004.2 XP_011539306.1 6 - 7 439 100
XM_017000652.1 XP_016856141.1 6 - 7 445 100
XM_017000653.1 XP_016856142.1 7 - 8 565 100
XM_017000654.1 XP_016856143.1 5 - 6 402 100
XM_017000655.1 XP_016856144.1 6 - 7 558 100
XM_017000656.1 XP_016856145.1 6 - 7 443 100
XM_017000657.1 XP_016856146.1 5 - 6 435 100
XM_017000658.1 XP_016856147.1 4 - 5 322 100
XM_017000659.1 XP_016856148.1 4 - 5 319 100
GenBank mRNA AK289805.1 - 5 - 6 364 100
AK295745.1 - 5 - 6 399 100
AK298129.1 - 4 - 5 322 100
AK303664.1 - 4 - 5 319 100
AL603098.1 - 5 - 6 388 100
BC029500.1 - 5 - 6 365 100
BC041667.1 - 5 - 6 332 100
BX648945.1 - 5 - 6 374 100
DC416048.1 - 5 - 6 405 100

View Full Product Details