Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- EYA3
- Entrez Gene ID
-
2140
- Gene Name
- EYA transcriptional coactivator and phosphatase 3
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 27970344 - 28088671 on Build GRCh38
View Full Product Details