The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_174897.2 | NP_777557.1 | 8 - 9 | 852 | 77 |
XM_017027663.1 | XP_016883152.1 | 8 - 9 | 900 | 77 | |
GenBank mRNA | AF465767.1 | - | 8 - 9 | 852 | 77 |