Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 243304594-243304867 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDCCAG8
Entrez Gene ID
10806
Gene Name
serologically defined colon cancer antigen 8
Gene Aliases
BBS16, CCCAP, CCCAP SLSN7, HSPC085, NPHP10, NY-CO-8, SLSN7, hCCCAP
UniGene
Hs.591530
Gene Chromosome Location
Chr.1: 243255328-243500092 on Build GRCh38
Transcripts
NM_006642.3 XM_005273013.4 XM_005273018.1 XM_005273021.4 XM_005273022.3 XM_005273023.4 XM_006711727.3 XM_006711728.3 XM_011544021.2 XM_011544022.2 XM_011544023.2 XM_011544024.2 XM_011544026.2 XM_011544028.2 XM_011544029.2 XM_017000103.1 XM_017000104.1 XM_017000105.1 XM_017000106.1 XM_017000107.1 XM_017000108.1 XM_017000109.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6460279 c.728T>A p.L243* Chr.1: 243304765-243304765 on Build GRCh38
2050845 c.712A>T p.I238F Chr.1: 243304749-243304749 on Build GRCh38
2050846 c.713T>A p.I238N Chr.1: 243304750-243304750 on Build GRCh38
6460281 c.728T>G p.L243W Chr.1: 243304765-243304765 on Build GRCh38
5613959 c.700G>T p.E234* Chr.1: 243304737-243304737 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757356055 Chr.1: 243304783-243304783 on Build GRCh38
rs370951398 Chr.1: 243304681-243304681 on Build GRCh38
rs758623782 Chr.1: 243304792-243304792 on Build GRCh38
rs753200349 Chr.1: 243304688-243304688 on Build GRCh38
rs753851659 Chr.1: 243304696-243304697 on Build GRCh38
rs146474700 Chr.1: 243304656-243304656 on Build GRCh38
rs768937029 Chr.1: 243304666-243304666 on Build GRCh38
rs937991135 Chr.1: 243304627-243304627 on Build GRCh38
rs373462087 Chr.1: 243304791-243304791 on Build GRCh38
rs773167463 Chr.1: 243304807-243304807 on Build GRCh38
rs777679135 Chr.1: 243304704-243304704 on Build GRCh38
rs767930611 Chr.1: 243304759-243304759 on Build GRCh38
rs923218609 Chr.1: 243304762-243304762 on Build GRCh38
rs762499427 Chr.1: 243304742-243304742 on Build GRCh38
rs764592655 Chr.1: 243304663-243304663 on Build GRCh38
rs7538434 Chr.1: 243304620-243304620 on Build GRCh38
rs200993724 Chr.1: 243304677-243304677 on Build GRCh38
rs186314584 Chr.1: 243304625-243304625 on Build GRCh38
rs747968552 Chr.1: 243304732-243304732 on Build GRCh38
rs756652195 Chr.1: 243304701-243304701 on Build GRCh38
rs751303224 Chr.1: 243304675-243304675 on Build GRCh38
rs570077019 Chr.1: 243304763-243304763 on Build GRCh38
rs996479713 Chr.1: 243304833-243304833 on Build GRCh38
rs772544112 Chr.1: 243304733-243304733 on Build GRCh38
rs768424996 Chr.1: 243304719-243304719 on Build GRCh38
rs1010240258 Chr.1: 243304630-243304630 on Build GRCh38
rs759278534 Chr.1: 243304758-243304758 on Build GRCh38
rs377544552 Chr.1: 243304804-243304804 on Build GRCh38
rs747993934 Chr.1: 243304802-243304802 on Build GRCh38
rs1055052228 Chr.1: 243304646-243304646 on Build GRCh38
rs756533597 Chr.1: 243304764-243304764 on Build GRCh38
rs757312997 Chr.1: 243304700-243304702 on Build GRCh38
rs377256554 Chr.1: 243304709-243304709 on Build GRCh38
rs749248291 Chr.1: 243304824-243304824 on Build GRCh38
rs771569101 Chr.1: 243304691-243304693 on Build GRCh38
rs762469561 Chr.1: 243304668-243304668 on Build GRCh38
rs559252091 Chr.1: 243304710-243304710 on Build GRCh38
rs761866388 Chr.1: 243304662-243304662 on Build GRCh38
rs749408172 Chr.1: 243304707-243304707 on Build GRCh38
rs1047427016 Chr.1: 243304788-243304788 on Build GRCh38
rs367814956 Chr.1: 243304670-243304670 on Build GRCh38
rs267607031 Chr.1: 243304716-243304716 on Build GRCh38
rs369949556 Chr.1: 243304801-243304801 on Build GRCh38
rs772782483 Chr.1: 243304667-243304667 on Build GRCh38
rs779020478 Chr.1: 243304790-243304790 on Build GRCh38
rs774139674 Chr.1: 243304755-243304755 on Build GRCh38
rs538969565 Chr.1: 243304687-243304687 on Build GRCh38
rs754344258 Chr.1: 243304768-243304768 on Build GRCh38
rs199629519 Chr.1: 243304766-243304766 on Build GRCh38
rs902664548 Chr.1: 243304737-243304737 on Build GRCh38
rs776422346 Chr.1: 243304720-243304720 on Build GRCh38
rs766024310 Chr.1: 243304747-243304747 on Build GRCh38
rs755359940 Chr.1: 243304676-243304676 on Build GRCh38

View Full Product Details