Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 21294570-21294835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPGRIP1
Entrez Gene ID
57096
Gene Name
retinitis pigmentosa GTPase regulator interacting protein 1
Gene Aliases
CORD13, LCA6, RGI1, RGRIP, RPGRIP, RPGRIP1d
UniGene
Hs.126035
Gene Chromosome Location
Chr.14: 21287977-21351316 on Build GRCh38
Transcripts
NM_020366.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
954370 c.160C>T p.R54C Chr.14: 21294751-21294751 on Build GRCh38
6200215 c.156A>C p.R52R Chr.14: 21294747-21294747 on Build GRCh38
5671077 c.126C>A p.N42K Chr.14: 21294717-21294717 on Build GRCh38
344273 c.191C>T p.S64F Chr.14: 21294782-21294782 on Build GRCh38
6075357 c.110C>T p.P37L Chr.14: 21294701-21294701 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773253797 Chr.14: 21294652-21294652 on Build GRCh38
rs199606938 Chr.14: 21294744-21294744 on Build GRCh38
rs754849037 Chr.14: 21294755-21294755 on Build GRCh38
rs941064555 Chr.14: 21294632-21294632 on Build GRCh38
rs752476122 Chr.14: 21294776-21294776 on Build GRCh38
rs929752463 Chr.14: 21294598-21294598 on Build GRCh38
rs1039495391 Chr.14: 21294637-21294637 on Build GRCh38
rs757352536 Chr.14: 21294798-21294798 on Build GRCh38
rs749726742 Chr.14: 21294639-21294640 on Build GRCh38
rs781479616 Chr.14: 21294639-21294639 on Build GRCh38
rs62646878 Chr.14: 21294786-21294786 on Build GRCh38
rs755845899 Chr.14: 21294778-21294778 on Build GRCh38
rs376250340 Chr.14: 21294752-21294752 on Build GRCh38
rs376435824 Chr.14: 21294707-21294707 on Build GRCh38
rs749473223 Chr.14: 21294795-21294795 on Build GRCh38
rs749943816 Chr.14: 21294701-21294701 on Build GRCh38
rs137853124 Chr.14: 21294785-21294785 on Build GRCh38
rs374171080 Chr.14: 21294746-21294746 on Build GRCh38
rs765765074 Chr.14: 21294753-21294753 on Build GRCh38
rs745852826 Chr.14: 21294627-21294627 on Build GRCh38
rs758376953 Chr.14: 21294628-21294628 on Build GRCh38
rs192003551 Chr.14: 21294745-21294745 on Build GRCh38
rs1046296429 Chr.14: 21294682-21294682 on Build GRCh38
rs771414528 Chr.14: 21294674-21294674 on Build GRCh38
rs200968268 Chr.14: 21294676-21294676 on Build GRCh38
rs1049489169 Chr.14: 21294601-21294601 on Build GRCh38
rs780057813 Chr.14: 21294633-21294633 on Build GRCh38
rs890848748 Chr.14: 21294621-21294621 on Build GRCh38
rs368952644 Chr.14: 21294648-21294648 on Build GRCh38
rs769885197 Chr.14: 21294651-21294651 on Build GRCh38
rs767737125 Chr.14: 21294690-21294690 on Build GRCh38
rs976713441 Chr.14: 21294775-21294775 on Build GRCh38
rs770933786 Chr.14: 21294732-21294732 on Build GRCh38
rs746804482 Chr.14: 21294635-21294635 on Build GRCh38
rs777888377 Chr.14: 21294727-21294727 on Build GRCh38
rs200510462 Chr.14: 21294686-21294686 on Build GRCh38
rs530008238 Chr.14: 21294754-21294754 on Build GRCh38
rs753553043 Chr.14: 21294692-21294692 on Build GRCh38
rs200285649 Chr.14: 21294636-21294636 on Build GRCh38
rs751975588 Chr.14: 21294718-21294718 on Build GRCh38
rs761540112 Chr.14: 21294695-21294695 on Build GRCh38
rs758465870 Chr.14: 21294702-21294702 on Build GRCh38
rs777977490 Chr.14: 21294780-21294780 on Build GRCh38
rs113468814 Chr.14: 21294613-21294613 on Build GRCh38
rs372867069 Chr.14: 21294657-21294657 on Build GRCh38
rs774956493 Chr.14: 21294736-21294736 on Build GRCh38
rs764967308 Chr.14: 21294748-21294748 on Build GRCh38
rs189509077 Chr.14: 21294698-21294698 on Build GRCh38
rs371044065 Chr.14: 21294728-21294728 on Build GRCh38
rs781081217 Chr.14: 21294761-21294761 on Build GRCh38

View Full Product Details