Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 34235794-34236045 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC12A6
Entrez Gene ID
9990
Gene Name
solute carrier family 12 member 6
Gene Aliases
ACCPN, KCC3, KCC3A, KCC3B
UniGene
Hs.510939
Gene Chromosome Location
Chr.15: 34229996-34338064 on Build GRCh38
Transcripts
NM_001042494.1 NM_001042495.1 NM_001042496.1 NM_001042497.1 NM_005135.2 NM_133647.1 XM_006720793.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3500532 c.3173G>A p.G1058E Chr.15: 34236042-34236042 on Build GRCh38
5951350 c.3182+1G>A p.? Chr.15: 34236014-34236014 on Build GRCh38
3500533 c.3155G>A p.G1052E Chr.15: 34236042-34236042 on Build GRCh38
4054112 c.3171A>G p.E1057E Chr.15: 34236044-34236044 on Build GRCh38
4054113 c.3153A>G p.E1051E Chr.15: 34236044-34236044 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs12439913 Chr.15: 34235854-34235854 on Build GRCh38
rs769675350 Chr.15: 34235975-34235975 on Build GRCh38
rs572445360 Chr.15: 34235903-34235903 on Build GRCh38
rs746008510 Chr.15: 34235964-34235965 on Build GRCh38
rs1029377802 Chr.15: 34235942-34235942 on Build GRCh38
rs1025443515 Chr.15: 34235828-34235828 on Build GRCh38
rs1057516969 Chr.15: 34236014-34236014 on Build GRCh38
rs545980432 Chr.15: 34235964-34235964 on Build GRCh38
rs185378817 Chr.15: 34235980-34235980 on Build GRCh38
rs536175598 Chr.15: 34235829-34235829 on Build GRCh38
rs1017967130 Chr.15: 34235889-34235889 on Build GRCh38
rs151271553 Chr.15: 34235996-34235996 on Build GRCh38
rs1005873410 Chr.15: 34235867-34235867 on Build GRCh38
rs1057517289 Chr.15: 34236021-34236022 on Build GRCh38
rs575233554 Chr.15: 34235827-34235827 on Build GRCh38
rs572443775 Chr.15: 34235904-34235904 on Build GRCh38
rs952646005 Chr.15: 34235837-34235837 on Build GRCh38
rs373473176 Chr.15: 34235972-34235972 on Build GRCh38
rs549730258 Chr.15: 34235894-34235894 on Build GRCh38
rs759622877 Chr.15: 34236011-34236011 on Build GRCh38
rs766658558 Chr.15: 34235983-34235983 on Build GRCh38
rs756712136 Chr.15: 34236022-34236022 on Build GRCh38
rs767931115 Chr.15: 34236018-34236018 on Build GRCh38
rs369219695 Chr.15: 34235971-34235971 on Build GRCh38
rs976420294 Chr.15: 34235940-34235940 on Build GRCh38
rs964030129 Chr.15: 34235898-34235898 on Build GRCh38
rs773298956 Chr.15: 34235977-34235977 on Build GRCh38
rs752965878 Chr.15: 34236019-34236019 on Build GRCh38

View Full Product Details