Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COPS8
Entrez Gene ID
10920
Gene Name
COP9 signalosome subunit 8
Gene Aliases
COP9, CSN8, SGN8
Chromosome Location
Chr.2: 237085441 - 237098846 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006710.4 NP_006701.1 4 - 5 855 71
NM_198189.2 NP_937832.1 5 - 6 920 71
GenBank mRNA AK289891.1 - 5 - 6 454 71
AK313437.1 - 4 - 5 398 71
BC003090.1 - 4 - 5 379 71
BC036499.1 - 4 - 5 989 71
BC080617.1 - 4 - 5 351 71
BG721393.1 - 5 - 6 458 71
CR456994.1 - 4 - 5 331 71
U51205.1 - 4 - 5 380 71

View Full Product Details