The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317804.1 | NP_001304733.1 | 9 - 10 | 927 | 120 |
NM_001317805.1 | NP_001304734.1 | 8 - 9 | 1442 | 120 | |
NM_014294.5 | NP_055109.1 | 8 - 9 | 1010 | 120 | |
XM_017013266.1 | XP_016868755.1 | 7 - 8 | 882 | 120 | |
XM_017013267.1 | XP_016868756.1 | 7 - 8 | 1226 | 120 | |
GenBank mRNA | AK289397.1 | - | 8 - 9 | 870 | 120 |
AK292777.1 | - | 8 - 9 | 794 | 120 | |
AK303411.1 | - | 9 - 10 | 855 | 120 | |
BC000687.2 | - | 8 - 9 | 832 | 120 | |
BC020283.1 | - | 8 - 9 | 825 | 120 | |
BC037738.1 | - | 8 - 9 | 922 | 120 | |
BT007359.1 | - | 8 - 9 | 750 | 120 | |
CR541654.1 | - | 8 - 9 | 750 | 120 | |
CR541739.1 | - | 8 - 9 | 750 | 120 | |
X63679.1 | - | 8 - 9 | 871 | 120 |