Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARSG
Entrez Gene ID
22901
Gene Name
arylsulfatase G
Gene Aliases
-
Chromosome Location
Chr.17: 68259089 - 68452000 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267727.1 NP_001254656.1 3 - 4 1027 55
NM_014960.4 NP_055775.2 3 - 4 1198 55
XM_005257170.3 XP_005257227.1 3 - 4 1211 55
XM_005257172.3 XP_005257229.1 3 - 4 1260 55
XM_006721777.3 XP_006721840.2 3 - 4 1225 55
XM_006721779.3 XP_006721842.1 3 - 4 1252 55
XM_011524535.2 XP_011522837.1 3 - 4 1223 55
XM_011524536.2 XP_011522838.1 3 - 4 1053 55
XM_011524537.1 XP_011522839.1 3 - 4 974 55
XM_011524538.2 XP_011522840.1 3 - 4 1225 55
XM_011524540.2 XP_011522842.1 3 - 4 1248 55
XM_011524541.2 XP_011522843.1 3 - 4 1291 55
XM_011524542.2 XP_011522844.1 3 - 4 1291 55
XM_011524543.2 XP_011522845.1 3 - 4 1291 55
XM_011524544.2 XP_011522846.1 3 - 4 1291 55
XM_011524545.2 XP_011522847.1 3 - 4 1249 55
XM_017024360.1 XP_016879849.1 3 - 4 1063 55
XM_017024361.1 XP_016879850.1 3 - 4 1211 55
XM_017024362.1 XP_016879851.1 3 - 4 1150 55
XM_017024363.1 XP_016879852.1 3 - 4 1056 55
XM_017024364.1 XP_016879853.1 3 - 4 1052 55
XM_017024365.1 XP_016879854.1 3 - 4 1040 55
XM_017024366.1 XP_016879855.1 3 - 4 1251 55
XM_017024367.1 XP_016879856.1 3 - 4 1053 55
XM_017024368.1 XP_016879857.1 3 - 4 1053 55
GenBank mRNA AB023218.1 - 2 - 3 860 55
AY358380.1 - 3 - 4 1198 55
BC012375.1 - 2 - 3 441 55

View Full Product Details