Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARSG
Entrez Gene ID
22901
Gene Name
arylsulfatase G
Gene Aliases
-
Chromosome Location
Chr.17: 68259089 - 68452000 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267727.1 NP_001254656.1 9 - 10 1718 82
NM_014960.4 NP_055775.2 9 - 10 1889 82
XM_005257170.3 XP_005257227.1 9 - 10 1902 82
XM_006721777.3 XP_006721840.2 9 - 10 1916 82
XM_011524535.2 XP_011522837.1 9 - 10 1914 82
XM_011524536.2 XP_011522838.1 9 - 10 1744 82
XM_011524537.1 XP_011522839.1 9 - 10 1665 82
XM_011524538.2 XP_011522840.1 9 - 10 1916 82
XM_011524540.2 XP_011522842.1 9 - 10 1936 82
XM_011524541.2 XP_011522843.1 9 - 10 1982 82
XM_011524542.2 XP_011522844.1 9 - 10 1982 82
XM_011524546.2 XP_011522848.1 4 - 5 587 82
XM_017024360.1 XP_016879849.1 9 - 10 1754 82
XM_017024361.1 XP_016879850.1 9 - 10 1902 82
XM_017024362.1 XP_016879851.1 9 - 10 1841 82
XM_017024363.1 XP_016879852.1 9 - 10 1747 82
XM_017024364.1 XP_016879853.1 9 - 10 1743 82
XM_017024365.1 XP_016879854.1 9 - 10 1731 82
XM_017024366.1 XP_016879855.1 9 - 10 1942 82
XM_017024367.1 XP_016879856.1 9 - 10 1744 82
GenBank mRNA AB023218.1 - 8 - 9 1551 82
AY358380.1 - 9 - 10 1889 82
BC012375.1 - 8 - 9 1132 82

View Full Product Details