Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38112332-38112587 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLA2G6
Entrez Gene ID
8398
Gene Name
phospholipase A2 group VI
Gene Aliases
CaI-PLA2, GVI, INAD1, IPLA2-VIA, NBIA2, NBIA2A, NBIA2B, PARK14, PLA2, PNPLA9, iPLA2, iPLA2beta
UniGene
Hs.170479
Gene Chromosome Location
Chr.22: 38111495-38192099 on Build GRCh38
Transcripts
NM_001004426.1 NM_001199562.1 NM_003560.2 XM_005261764.2 XM_005261765.1 XM_005261766.1 XM_006724332.3 XM_011530422.1 XM_011530423.1 XM_011530424.1 XM_011530425.1 XM_017028981.1 XM_017028982.1 XM_017028983.1 XM_017028984.1 XM_017028985.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4388868 c.2212C>A p.P738T Chr.22: 38112568-38112568 on Build GRCh38
6499871 c.2265C>G p.I755M Chr.22: 38112515-38112515 on Build GRCh38
7384180 c.2240G>A p.R747Q Chr.22: 38112540-38112540 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs374303522 Chr.22: 38112474-38112474 on Build GRCh38
rs963927447 Chr.22: 38112520-38112520 on Build GRCh38
rs746013568 Chr.22: 38112562-38112562 on Build GRCh38
rs548439730 Chr.22: 38112563-38112563 on Build GRCh38
rs761294759 Chr.22: 38112491-38112491 on Build GRCh38
rs375779632 Chr.22: 38112472-38112472 on Build GRCh38
rs121908687 Chr.22: 38112541-38112541 on Build GRCh38
rs986738833 Chr.22: 38112481-38112481 on Build GRCh38
rs530348521 Chr.22: 38112559-38112559 on Build GRCh38
rs11570766 Chr.22: 38112372-38112372 on Build GRCh38
rs1043504064 Chr.22: 38112399-38112399 on Build GRCh38
rs587784351 Chr.22: 38112534-38112534 on Build GRCh38
rs563222818 Chr.22: 38112544-38112544 on Build GRCh38
rs796331166 Chr.22: 38112405-38112405 on Build GRCh38
rs993482020 Chr.22: 38112468-38112468 on Build GRCh38
rs749437245 Chr.22: 38112462-38112462 on Build GRCh38
rs924915026 Chr.22: 38112404-38112404 on Build GRCh38
rs544263173 Chr.22: 38112527-38112527 on Build GRCh38
rs372267803 Chr.22: 38112490-38112490 on Build GRCh38
rs955215757 Chr.22: 38112351-38112351 on Build GRCh38
rs554827625 Chr.22: 38112455-38112455 on Build GRCh38
rs914530056 Chr.22: 38112402-38112402 on Build GRCh38
rs11570765 Chr.22: 38112434-38112434 on Build GRCh38
rs1045677686 Chr.22: 38112452-38112452 on Build GRCh38
rs34910119 Chr.22: 38112371-38112372 on Build GRCh38
rs587784349 Chr.22: 38112565-38112565 on Build GRCh38
rs368514303 Chr.22: 38112546-38112546 on Build GRCh38
rs571458275 Chr.22: 38112457-38112457 on Build GRCh38
rs11570764 Chr.22: 38112461-38112461 on Build GRCh38
rs577215031 Chr.22: 38112487-38112487 on Build GRCh38
rs182038896 Chr.22: 38112415-38112415 on Build GRCh38
rs368835758 Chr.22: 38112489-38112489 on Build GRCh38
rs886184330 Chr.22: 38112449-38112449 on Build GRCh38
rs41274545 Chr.22: 38112361-38112361 on Build GRCh38
rs121908686 Chr.22: 38112558-38112558 on Build GRCh38
rs764789953 Chr.22: 38112519-38112519 on Build GRCh38
rs949569745 Chr.22: 38112447-38112447 on Build GRCh38
rs938490717 Chr.22: 38112443-38112443 on Build GRCh38
rs751225193 Chr.22: 38112531-38112531 on Build GRCh38
rs753256678 Chr.22: 38112375-38112375 on Build GRCh38
rs754070657 Chr.22: 38112358-38112358 on Build GRCh38
rs779749576 Chr.22: 38112530-38112530 on Build GRCh38
rs373689745 Chr.22: 38112458-38112458 on Build GRCh38
rs565332748 Chr.22: 38112467-38112467 on Build GRCh38
rs759942729 Chr.22: 38112485-38112485 on Build GRCh38
rs370465424 Chr.22: 38112471-38112471 on Build GRCh38
rs749966284 Chr.22: 38112521-38112521 on Build GRCh38
rs376144077 Chr.22: 38112540-38112540 on Build GRCh38
rs587784350 Chr.22: 38112547-38112547 on Build GRCh38
rs936682716 Chr.22: 38112388-38112388 on Build GRCh38

View Full Product Details