Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 202946384-202946655 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADIPOR1
Entrez Gene ID
51094
Gene Name
adiponectin receptor 1
Gene Aliases
ACDCR1, CGI-45, CGI45, PAQR1, TESBP1A
UniGene
Hs.5298
Gene Chromosome Location
Chr.1: 202940825-202958572 on Build GRCh38
Transcripts
NM_001290553.1 NM_001290557.1 NM_001290629.1 NM_015999.5 XM_017001407.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5978207 c.389G>A p.R130H Chr.1: 202946480-202946480 on Build GRCh38
242856 c.326A>G p.Y109C Chr.1: 202946543-202946543 on Build GRCh38
6573773 c.295G>C p.V99L Chr.1: 202946574-202946574 on Build GRCh38
7263081 c.325T>C p.Y109H Chr.1: 202946544-202946544 on Build GRCh38
242855 c.423T>C p.H141H Chr.1: 202946446-202946446 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764226232 Chr.1: 202946505-202946505 on Build GRCh38
rs889809506 Chr.1: 202946488-202946488 on Build GRCh38
rs773450844 Chr.1: 202946530-202946530 on Build GRCh38
rs1030321794 Chr.1: 202946458-202946458 on Build GRCh38
rs956441099 Chr.1: 202946480-202946480 on Build GRCh38
rs1008227340 Chr.1: 202946501-202946501 on Build GRCh38
rs1019743252 Chr.1: 202946536-202946536 on Build GRCh38
rs200676846 Chr.1: 202946597-202946597 on Build GRCh38
rs760488217 Chr.1: 202946440-202946440 on Build GRCh38
rs377163409 Chr.1: 202946548-202946548 on Build GRCh38
rs761522938 Chr.1: 202946423-202946423 on Build GRCh38
rs765136165 Chr.1: 202946624-202946624 on Build GRCh38
rs1043213 Chr.1: 202946476-202946476 on Build GRCh38
rs760402203 Chr.1: 202946610-202946610 on Build GRCh38
rs757529476 Chr.1: 202946518-202946518 on Build GRCh38
rs141706866 Chr.1: 202946473-202946473 on Build GRCh38
rs781585434 Chr.1: 202946523-202946523 on Build GRCh38
rs2275735 Chr.1: 202946427-202946427 on Build GRCh38
rs766179380 Chr.1: 202946613-202946613 on Build GRCh38
rs773885338 Chr.1: 202946566-202946566 on Build GRCh38
rs759481683 Chr.1: 202946621-202946621 on Build GRCh38
rs753825645 Chr.1: 202946449-202946449 on Build GRCh38
rs967783120 Chr.1: 202946541-202946541 on Build GRCh38
rs994211930 Chr.1: 202946576-202946576 on Build GRCh38
rs377378130 Chr.1: 202946581-202946581 on Build GRCh38
rs746193003 Chr.1: 202946527-202946527 on Build GRCh38
rs766267373 Chr.1: 202946441-202946441 on Build GRCh38
rs749789403 Chr.1: 202946547-202946547 on Build GRCh38
rs773250168 Chr.1: 202946437-202946438 on Build GRCh38
rs7518457 Chr.1: 202946408-202946408 on Build GRCh38
rs773792891 Chr.1: 202946421-202946421 on Build GRCh38
rs751626519 Chr.1: 202946516-202946516 on Build GRCh38
rs767202058 Chr.1: 202946426-202946426 on Build GRCh38
rs768227600 Chr.1: 202946418-202946418 on Build GRCh38
rs754925881 Chr.1: 202946481-202946481 on Build GRCh38
rs141511034 Chr.1: 202946582-202946582 on Build GRCh38
rs776353126 Chr.1: 202946616-202946616 on Build GRCh38
rs899982679 Chr.1: 202946428-202946428 on Build GRCh38
rs772885058 Chr.1: 202946602-202946602 on Build GRCh38

View Full Product Details