Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 58431936-58432209 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIPC
Entrez Gene ID
3990
Gene Name
lipase C, hepatic type
Gene Aliases
HDLCQ12, HL, HTGL, LIPH
UniGene
Hs.654472
Gene Chromosome Location
Chr.15: 58410754-58568952 on Build GRCh38
Transcripts
NM_000236.2 XM_005254372.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
347549 c.59C>G p.S20* Chr.15: 58432091-58432091 on Build GRCh38
963334 c.48C>T p.I16I Chr.15: 58432080-58432080 on Build GRCh38
4698127 c.64G>A p.A22T Chr.15: 58432096-58432096 on Build GRCh38
4698126 c.10A>T p.S4C Chr.15: 58432042-58432042 on Build GRCh38
4055890 c.8C>T p.T3I Chr.15: 58432040-58432040 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370723304 Chr.15: 58432010-58432010 on Build GRCh38
rs371941209 Chr.15: 58431966-58431966 on Build GRCh38
rs756169441 Chr.15: 58432036-58432036 on Build GRCh38
rs137926228 Chr.15: 58432128-58432128 on Build GRCh38
rs780267766 Chr.15: 58432110-58432110 on Build GRCh38
rs767495112 Chr.15: 58431996-58431996 on Build GRCh38
rs775421240 Chr.15: 58432007-58432007 on Build GRCh38
rs755550316 Chr.15: 58432027-58432027 on Build GRCh38
rs555484619 Chr.15: 58431981-58431981 on Build GRCh38
rs185851272 Chr.15: 58432148-58432148 on Build GRCh38
rs959504349 Chr.15: 58432083-58432083 on Build GRCh38
rs770649146 Chr.15: 58432046-58432046 on Build GRCh38
rs753178180 Chr.15: 58432135-58432135 on Build GRCh38
rs868759996 Chr.15: 58432132-58432132 on Build GRCh38
rs756402865 Chr.15: 58432037-58432037 on Build GRCh38
rs772192757 Chr.15: 58432087-58432087 on Build GRCh38
rs958264527 Chr.15: 58431978-58431978 on Build GRCh38
rs544198137 Chr.15: 58432038-58432038 on Build GRCh38
rs745366705 Chr.15: 58432079-58432079 on Build GRCh38
rs750577708 Chr.15: 58432014-58432014 on Build GRCh38
rs377671394 Chr.15: 58432150-58432150 on Build GRCh38
rs201219858 Chr.15: 58432125-58432125 on Build GRCh38
rs139328592 Chr.15: 58432049-58432049 on Build GRCh38
rs778694967 Chr.15: 58432166-58432166 on Build GRCh38
rs376602263 Chr.15: 58432154-58432154 on Build GRCh38
rs369107346 Chr.15: 58432094-58432094 on Build GRCh38
rs11631342 Chr.15: 58432184-58432184 on Build GRCh38
rs751699159 Chr.15: 58432124-58432124 on Build GRCh38
rs999971544 Chr.15: 58432179-58432179 on Build GRCh38
rs933615635 Chr.15: 58432167-58432167 on Build GRCh38
rs374183304 Chr.15: 58432146-58432146 on Build GRCh38
rs760809626 Chr.15: 58432040-58432041 on Build GRCh38
rs775738121 Chr.15: 58432031-58432038 on Build GRCh38
rs149520038 Chr.15: 58432173-58432173 on Build GRCh38
rs751574296 Chr.15: 58432026-58432026 on Build GRCh38
rs1032529780 Chr.15: 58431960-58431960 on Build GRCh38
rs776432016 Chr.15: 58432103-58432103 on Build GRCh38
rs769006323 Chr.15: 58432045-58432046 on Build GRCh38
rs201654179 Chr.15: 58432085-58432085 on Build GRCh38
rs745901678 Chr.15: 58432008-58432008 on Build GRCh38
rs372381397 Chr.15: 58432028-58432028 on Build GRCh38
rs372805598 Chr.15: 58432018-58432018 on Build GRCh38
rs145398861 Chr.15: 58432099-58432099 on Build GRCh38
rs926671529 Chr.15: 58432042-58432042 on Build GRCh38
rs748972683 Chr.15: 58432041-58432041 on Build GRCh38
rs760554069 Chr.15: 58432098-58432098 on Build GRCh38
rs8192701 Chr.15: 58432172-58432172 on Build GRCh38
rs374467536 Chr.15: 58432017-58432017 on Build GRCh38

View Full Product Details