Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 25911730-25911999 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APP
Entrez Gene ID
351
Gene Name
amyloid beta precursor protein
Gene Aliases
AAA, ABETA, ABPP, AD1, APPI, CTFgamma, CVAP, PN-II, PN2
UniGene
Hs.434980
Gene Chromosome Location
Chr.21: 25880550-26171128 on Build GRCh38
Transcripts
NM_000484.3 NM_001136016.3 NM_001136129.2 NM_001136130.2 NM_001136131.2 NM_001204301.1 NM_001204302.1 NM_001204303.1 NM_201413.2 NM_201414.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7177135 c.1795G>C p.E599Q Chr.21: 25911855-25911855 on Build GRCh38
5392745 c.1864C>T p.H622Y Chr.21: 25911786-25911786 on Build GRCh38
6018032 c.1818C>T p.L606L Chr.21: 25911832-25911832 on Build GRCh38
1615936 c.1794C>T p.T598T Chr.21: 25911856-25911856 on Build GRCh38
2841342 c.1800G>A p.T600T Chr.21: 25911850-25911850 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748980864 Chr.21: 25911836-25911836 on Build GRCh38
rs201874897 Chr.21: 25911879-25911879 on Build GRCh38
rs759926843 Chr.21: 25911791-25911791 on Build GRCh38
rs200769792 Chr.21: 25911924-25911924 on Build GRCh38
rs748858141 Chr.21: 25911900-25911900 on Build GRCh38
rs150151009 Chr.21: 25911841-25911841 on Build GRCh38
rs763057681 Chr.21: 25911950-25911950 on Build GRCh38
rs755596937 Chr.21: 25911835-25911835 on Build GRCh38
rs145371658 Chr.21: 25911899-25911899 on Build GRCh38
rs199887707 Chr.21: 25911840-25911840 on Build GRCh38
rs138434918 Chr.21: 25911850-25911850 on Build GRCh38
rs755645885 Chr.21: 25911885-25911885 on Build GRCh38
rs140304729 Chr.21: 25911855-25911855 on Build GRCh38
rs761864553 Chr.21: 25911807-25911807 on Build GRCh38
rs765301301 Chr.21: 25911860-25911860 on Build GRCh38
rs201794320 Chr.21: 25911928-25911928 on Build GRCh38
rs1038162399 Chr.21: 25911954-25911954 on Build GRCh38
rs769486659 Chr.21: 25911785-25911785 on Build GRCh38
rs201479601 Chr.21: 25911801-25911801 on Build GRCh38
rs773379255 Chr.21: 25911861-25911861 on Build GRCh38
rs778495527 Chr.21: 25911902-25911902 on Build GRCh38
rs780774625 Chr.21: 25911778-25911778 on Build GRCh38
rs771132182 Chr.21: 25911959-25911959 on Build GRCh38
rs751367779 Chr.21: 25911873-25911873 on Build GRCh38
rs201724379 Chr.21: 25911849-25911849 on Build GRCh38
rs111269011 Chr.21: 25911964-25911964 on Build GRCh38
rs758117754 Chr.21: 25911818-25911818 on Build GRCh38
rs200088099 Chr.21: 25911851-25911851 on Build GRCh38
rs200521782 Chr.21: 25911960-25911960 on Build GRCh38
rs200922667 Chr.21: 25911817-25911817 on Build GRCh38
rs374277868 Chr.21: 25911966-25911966 on Build GRCh38
rs756941168 Chr.21: 25911901-25911901 on Build GRCh38
rs201221707 Chr.21: 25911904-25911904 on Build GRCh38
rs374799888 Chr.21: 25911837-25911837 on Build GRCh38
rs199761619 Chr.21: 25911783-25911783 on Build GRCh38
rs763182419 Chr.21: 25911869-25911869 on Build GRCh38
rs201858248 Chr.21: 25911823-25911823 on Build GRCh38
rs766634145 Chr.21: 25911871-25911871 on Build GRCh38
rs200150248 Chr.21: 25911880-25911880 on Build GRCh38
rs201817181 Chr.21: 25911856-25911856 on Build GRCh38
rs773060369 Chr.21: 25911790-25911790 on Build GRCh38
rs781060683 Chr.21: 25911826-25911826 on Build GRCh38
rs137865262 Chr.21: 25911961-25911961 on Build GRCh38
rs764559840 Chr.21: 25911802-25911802 on Build GRCh38
rs533667466 Chr.21: 25911912-25911912 on Build GRCh38
rs747684890 Chr.21: 25911830-25911830 on Build GRCh38
rs776739799 Chr.21: 25911804-25911804 on Build GRCh38
rs933759872 Chr.21: 25911883-25911883 on Build GRCh38
rs200539821 Chr.21: 25911796-25911796 on Build GRCh38
rs754790092 Chr.21: 25911875-25911875 on Build GRCh38
rs541374184 Chr.21: 25911825-25911825 on Build GRCh38
rs773251655 Chr.21: 25911937-25911937 on Build GRCh38
rs199905452 Chr.21: 25911863-25911863 on Build GRCh38
rs201264568 Chr.21: 25911874-25911874 on Build GRCh38
rs201160106 Chr.21: 25911769-25911769 on Build GRCh38
rs748630651 Chr.21: 25911925-25911925 on Build GRCh38
rs112263157 Chr.21: 25911810-25911810 on Build GRCh38

View Full Product Details