Product Details

Assay Reference Genome
Location

Chr.21:34370749 on build GRCh38
Cytoband
21q22.11
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
9992
Gene Symbol
KCNE2
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 2
Gene Aliases
ATFB4, LQT5, LQT6, MIRP1
Location
Chr.21:34364024-34371141 on build GRCh38
Assay Gene Location
Within Exon 2
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
KCNE2 NM_172201.1 2 411 NP_751951.1
AF071002.1 1 344 AAD28086.1
AF302095.1 2 411 AAG13416.1
BC093892.1 2 362 AAH93892.1
BC112087.1 2 363 AAI12088.1
DQ192290.1 ABB00303.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv7824n54 Chr21:34352440 - 34535115 on Build GRCh38 Gain C21orf140 SMIM11A LOC105372793 KCNE1 LOC388820 KCNE2 RCAN1
dgv4426n100 Chr21:34339322 - 34534919 on Build GRCh38 Gain C21orf140 SMIM11A LOC105372793 KCNE1 LOC388820 KCNE2 RCAN1
nsv834088 Chr21:34241420 - 34379776 on Build GRCh38 Loss SMIM11A KCNE2
nsv1055429 Chr21:34299853 - 34516938 on Build GRCh38 Gain C21orf140 SMIM11A LOC105372793 KCNE1 LOC388820 KCNE2 RCAN1
esv3575386 Chr21:34353927 - 34534570 on Build GRCh38 Gain C21orf140 SMIM11A LOC105372793 KCNE1 LOC388820 KCNE2 RCAN1
esv2762109 Chr21:34350696 - 34531755 on Build GRCh38 Gain C21orf140 SMIM11A LOC105372793 KCNE1 LOC388820 KCNE2 RCAN1

View Full Product Details