Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 55151215-55151448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGFR
Entrez Gene ID
1956
Gene Name
epidermal growth factor receptor
Gene Aliases
ERBB, ERBB1, HER1, NISBD2, PIG61, mENA
UniGene
Hs.488293
Gene Chromosome Location
Chr.7: 55019032-55207338 on Build GRCh38
Transcripts
NM_005228.3 NM_201282.1 NM_201283.1 NM_201284.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5986342 c.628C>T p.L210L Chr.7: 55151362-55151362 on Build GRCh38
6921883 c.602_603insT p.A202fs*29 Chr.7: 55151336-55151337 on Build GRCh38
5407719 c.622C>T p.Q208* Chr.7: 55151356-55151356 on Build GRCh38
3721610 c.608G>A p.G203E Chr.7: 55151342-55151342 on Build GRCh38
6921885 c.602_603insT p.A202fs*29 Chr.7: 55151336-55151337 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778238294 Chr.7: 55151266-55151266 on Build GRCh38
rs747000271 Chr.7: 55151273-55151273 on Build GRCh38
rs773349388 Chr.7: 55151313-55151313 on Build GRCh38
rs776568205 Chr.7: 55151295-55151295 on Build GRCh38
rs769546191 Chr.7: 55151412-55151412 on Build GRCh38
rs551380760 Chr.7: 55151400-55151401 on Build GRCh38
rs377574069 Chr.7: 55151399-55151399 on Build GRCh38
rs770074413 Chr.7: 55151308-55151308 on Build GRCh38
rs756965461 Chr.7: 55151265-55151265 on Build GRCh38
rs747951029 Chr.7: 55151410-55151410 on Build GRCh38
rs780798595 Chr.7: 55151402-55151402 on Build GRCh38
rs766948894 Chr.7: 55151404-55151404 on Build GRCh38
rs746871680 Chr.7: 55151398-55151398 on Build GRCh38
rs753366090 Chr.7: 55151260-55151260 on Build GRCh38
rs748320322 Chr.7: 55151340-55151340 on Build GRCh38
rs377044664 Chr.7: 55151279-55151279 on Build GRCh38
rs757968655 Chr.7: 55151392-55151392 on Build GRCh38
rs764911170 Chr.7: 55151388-55151388 on Build GRCh38
rs773472875 Chr.7: 55151414-55151414 on Build GRCh38
rs766507267 Chr.7: 55151321-55151321 on Build GRCh38
rs573402976 Chr.7: 55151237-55151237 on Build GRCh38
rs963953337 Chr.7: 55151407-55151410 on Build GRCh38
rs370376501 Chr.7: 55151328-55151328 on Build GRCh38
rs767885569 Chr.7: 55151255-55151255 on Build GRCh38
rs753129657 Chr.7: 55151358-55151358 on Build GRCh38
rs369677373 Chr.7: 55151238-55151238 on Build GRCh38
rs771539767 Chr.7: 55151252-55151252 on Build GRCh38
rs751473624 Chr.7: 55151277-55151282 on Build GRCh38
rs534848277 Chr.7: 55151420-55151420 on Build GRCh38
rs748275089 Chr.7: 55151301-55151301 on Build GRCh38
rs763086693 Chr.7: 55151316-55151316 on Build GRCh38
rs756486534 Chr.7: 55151381-55151381 on Build GRCh38
rs368505460 Chr.7: 55151402-55151403 on Build GRCh38
rs757885405 Chr.7: 55151272-55151272 on Build GRCh38
rs768512891 Chr.7: 55151285-55151285 on Build GRCh38
rs779724563 Chr.7: 55151393-55151393 on Build GRCh38
rs954768311 Chr.7: 55151380-55151381 on Build GRCh38
rs745363899 Chr.7: 55151268-55151268 on Build GRCh38
rs767829342 Chr.7: 55151344-55151344 on Build GRCh38

View Full Product Details