Product Details

Assay Reference Genome
Location

Chr.X:18650470 on build GRCh38
Cytoband
Xp22.13
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
6792
Gene Symbol
CDKL5
Gene Name
cyclin dependent kinase like 5
Gene Aliases
CFAP247, EIEE2, ISSX, STK9
Location
Chr.X:18425605-18653629 on build GRCh38
Assay Gene Location
Within Exon 22
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
CDKL5 NM_001037343.1 21 3107 NP_001032420.1
NM_003159.2 20 3111 NP_003150.1
AI286150.1
AY217744.1 20 3079 AAO64440.1
Y15057.1 20 3079 CAA75342.1
Entrez Gene ID
6247
Gene Symbol
RS1
Gene Name
retinoschisin 1
Gene Aliases
RS, XLRS1
Location
Chr.X:18639688-18672103 on build GRCh38
Assay Gene Location
Within Intron 3
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
RS1 NM_000330.3 NP_000321.1
AF014459.1 AAC17928.1
AL049684.1 1 68
BQ636812.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv6821 ChrX:18622224 - 18656388 on Build GRCh38 Insertion RS1 CDKL5
nsv528654 ChrX:18627439 - 18660570 on Build GRCh38 Gain RS1 CDKL5

View Full Product Details