Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 58542471-58542716 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIPC
Entrez Gene ID
3990
Gene Name
lipase C, hepatic type
Gene Aliases
HDLCQ12, HL, HTGL, LIPH
UniGene
Hs.654472
Gene Chromosome Location
Chr.15: 58410754-58568952 on Build GRCh38
Transcripts
NM_000236.2 XM_005254372.1 XM_005254374.4 XM_006720502.3 XM_017022176.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
700861 c.476G>A p.R159Q Chr.15: 58542553-58542553 on Build GRCh38
6260786 c.552G>T p.T184T Chr.15: 58542629-58542629 on Build GRCh38
312530 c.496G>T p.G166W Chr.15: 58542573-58542573 on Build GRCh38
282437 c.552G>A p.T184T Chr.15: 58542629-58542629 on Build GRCh38
4166362 c.564G>C p.G188G Chr.15: 58542641-58542641 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761213115 Chr.15: 58542608-58542608 on Build GRCh38
rs770297325 Chr.15: 58542625-58542625 on Build GRCh38
rs554643312 Chr.15: 58542498-58542498 on Build GRCh38
rs775650887 Chr.15: 58542677-58542677 on Build GRCh38
rs375115836 Chr.15: 58542571-58542571 on Build GRCh38
rs770049039 Chr.15: 58542513-58542513 on Build GRCh38
rs774356974 Chr.15: 58542585-58542585 on Build GRCh38
rs765814048 Chr.15: 58542681-58542682 on Build GRCh38
rs200758590 Chr.15: 58542586-58542586 on Build GRCh38
rs200722587 Chr.15: 58542580-58542580 on Build GRCh38
rs781032963 Chr.15: 58542637-58542637 on Build GRCh38
rs145111892 Chr.15: 58542692-58542692 on Build GRCh38
rs767952446 Chr.15: 58542594-58542594 on Build GRCh38
rs776354069 Chr.15: 58542573-58542573 on Build GRCh38
rs768058337 Chr.15: 58542524-58542524 on Build GRCh38
rs376354109 Chr.15: 58542552-58542552 on Build GRCh38
rs759642964 Chr.15: 58542593-58542593 on Build GRCh38
rs556887747 Chr.15: 58542519-58542519 on Build GRCh38
rs764689519 Chr.15: 58542685-58542685 on Build GRCh38
rs543544840 Chr.15: 58542664-58542664 on Build GRCh38
rs753055565 Chr.15: 58542532-58542532 on Build GRCh38
rs771112217 Chr.15: 58542655-58542655 on Build GRCh38
rs761264458 Chr.15: 58542679-58542679 on Build GRCh38
rs753168442 Chr.15: 58542609-58542609 on Build GRCh38
rs373270467 Chr.15: 58542666-58542666 on Build GRCh38
rs768463883 Chr.15: 58542572-58542572 on Build GRCh38
rs369742319 Chr.15: 58542613-58542613 on Build GRCh38
rs762411833 Chr.15: 58542503-58542505 on Build GRCh38
rs142833050 Chr.15: 58542550-58542550 on Build GRCh38
rs568040695 Chr.15: 58542505-58542505 on Build GRCh38
rs138967170 Chr.15: 58542628-58542628 on Build GRCh38
rs543350478 Chr.15: 58542676-58542676 on Build GRCh38
rs756908602 Chr.15: 58542548-58542548 on Build GRCh38
rs750119964 Chr.15: 58542695-58542695 on Build GRCh38
rs145742763 Chr.15: 58542553-58542553 on Build GRCh38
rs188353743 Chr.15: 58542621-58542621 on Build GRCh38
rs756518358 Chr.15: 58542538-58542538 on Build GRCh38
rs754185467 Chr.15: 58542618-58542618 on Build GRCh38
rs752145667 Chr.15: 58542688-58542688 on Build GRCh38
rs375429385 Chr.15: 58542631-58542631 on Build GRCh38
rs924548064 Chr.15: 58542556-58542556 on Build GRCh38
rs760027937 Chr.15: 58542522-58542522 on Build GRCh38
rs765515473 Chr.15: 58542582-58542582 on Build GRCh38
rs572467817 Chr.15: 58542620-58542620 on Build GRCh38
rs560972096 Chr.15: 58542622-58542622 on Build GRCh38
rs370340255 Chr.15: 58542557-58542557 on Build GRCh38
rs690 Chr.15: 58542542-58542542 on Build GRCh38
rs763115101 Chr.15: 58542517-58542517 on Build GRCh38
rs746964093 Chr.15: 58542629-58542629 on Build GRCh38
rs746039268 Chr.15: 58542669-58542669 on Build GRCh38
rs776099493 Chr.15: 58542602-58542602 on Build GRCh38
rs771180270 Chr.15: 58542575-58542575 on Build GRCh38
rs745802629 Chr.15: 58542499-58542499 on Build GRCh38
rs773389548 Chr.15: 58542515-58542515 on Build GRCh38
rs762245376 Chr.15: 58542689-58542689 on Build GRCh38
rs760504900 Chr.15: 58542509-58542509 on Build GRCh38
rs778434866 Chr.15: 58542495-58542495 on Build GRCh38
rs770594015 Chr.15: 58542492-58542492 on Build GRCh38
rs6073 Chr.15: 58542518-58542518 on Build GRCh38
rs368820559 Chr.15: 58542645-58542645 on Build GRCh38
rs368130353 Chr.15: 58542574-58542574 on Build GRCh38

View Full Product Details