The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277061.1 | NP_001263990.1 | 7 - 8 | 1120 | 65 |
NM_020194.5 | NP_064579.3 | 6 - 7 | 857 | 65 | |
XM_005246681.3 | XP_005246738.1 | 7 - 8 | 1040 | 65 | |
XM_005246683.1 | XP_005246740.1 | 6 - 7 | 862 | 65 | |
XM_006712637.1 | XP_006712700.1 | 5 - 6 | 764 | 65 | |
XM_011511500.1 | XP_011509802.1 | 7 - 8 | 912 | 65 | |
XM_011511501.1 | XP_011509803.1 | 6 - 7 | 933 | 65 | |
GenBank mRNA | AF246239.1 | - | 6 - 7 | 857 | 65 |
AF258660.1 | - | 7 - 8 | 975 | 65 |