The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173794.3 | NP_776155.1 | 1 - 2 | 171 | 131 |
GenBank mRNA | BC035015.2 | - | 1 - 2 | 55 | 131 |
BC042813.1 | - | 1 - 2 | 166 | 131 | |
BM837991.1 | - | 1 - 2 | 197 | 131 |