The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_121682.1 | 1 - 1 | 1046 | 67 | |
GenBank mRNA | AK098456.1 | - | 1 - 1 | 1047 | 67 |
BC038738.1 | - | 1 - 1 | 749 | 67 |