The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005335.1 | NP_001005335.1 | 13 - 13 | 1720 | 144 |
NM_001533.2 | NP_001524.2 | 13 - 13 | 1954 | 144 | |
XM_011526886.1 | XP_011525188.1 | 13 - 13 | 1743 | 144 | |
XM_011526887.1 | XP_011525189.1 | 13 - 13 | 1787 | 144 | |
XM_011526888.2 | XP_011525190.1 | 11 - 11 | 1842 | 144 | |
XM_011526889.1 | XP_011525191.1 | 13 - 13 | 1838 | 144 | |
GenBank mRNA | AK094966.1 | - | 6 - 6 | 2942 | 147 |
AK097975.1 | - | 4 - 4 | 1959 | 144 | |
AV743120.2 | - | 1 - 1 | 228 | 144 | |
BC069184.1 | - | 13 - 13 | 1884 | 144 | |
BM997815.1 | - | 1 - 1 | 144 | ||
X16135.1 | - | 13 - 13 | 1873 | 144 |