The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308025.1 | NP_001294954.1 | 21 - 22 | 5604 | 106 |
NM_152259.3 | NP_689472.3 | 21 - 22 | 5607 | 106 | |
GenBank mRNA | AK074727.1 | - | 2 - 3 | 2099 | 106 |
AK123612.1 | - | 14 - 15 | 3707 | 106 | |
AK315750.1 | - | 2 - 3 | 1919 | 106 | |
BC002881.2 | - | 2 - 3 | 518 | 106 | |
BC033209.1 | - | 2 - 3 | 1649 | 106 | |
BX538335.1 | - | 8 - 9 | 2908 | 106 | |
GQ227787.1 | - | 21 - 22 | 5604 | 106 |