Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LYRM7
Entrez Gene ID
90624
Gene Name
LYR motif containing 7
Gene Aliases
C5orf31, MC3DN8, MZM1L
Chromosome Location
Chr.5: 131170914 - 131205426 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181705.3 NP_859056.2 4 - 5 345 110
GenBank mRNA AK000803.1 - 4 - 5 316 110
BC037930.1 - 4 - 5 297 110
BC047079.1 - 4 - 5 275 110
BI601105.1 - 4 - 5 321 110
DA996312.1 - 4 - 5 347 110

View Full Product Details