Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38126266-38126531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLA2G6
Entrez Gene ID
8398
Gene Name
phospholipase A2 group VI
Gene Aliases
CaI-PLA2, GVI, INAD1, IPLA2-VIA, NBIA2, NBIA2A, NBIA2B, PARK14, PLA2, PNPLA9, iPLA2, iPLA2beta
UniGene
Hs.170479
Gene Chromosome Location
Chr.22: 38111495-38192099 on Build GRCh38
Transcripts
NM_001004426.1 NM_001199562.1 NM_003560.2 XM_005261764.2 XM_005261765.1 XM_005261766.1 XM_006724332.3 XM_011530422.1 XM_011530423.1 XM_011530424.1 XM_011530425.1 XM_011530426.2 XM_017028981.1 XM_017028982.1 XM_017028983.1 XM_017028984.1 XM_017028985.1 XM_017028986.1 XM_017028987.1 XM_017028988.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5859358 c.1354C>T p.Q452* Chr.22: 38126444-38126444 on Build GRCh38
5698418 c.1384A>C p.K462Q Chr.22: 38126414-38126414 on Build GRCh38
1034104 c.1410G>A p.M470I Chr.22: 38126388-38126388 on Build GRCh38
7137890 c.1425G>A p.R475R Chr.22: 38126373-38126373 on Build GRCh38
7402167 c.1362C>T p.L454L Chr.22: 38126436-38126436 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764618392 Chr.22: 38126338-38126339 on Build GRCh38
rs770908303 Chr.22: 38126336-38126336 on Build GRCh38
rs557626802 Chr.22: 38126318-38126318 on Build GRCh38
rs775663600 Chr.22: 38126343-38126343 on Build GRCh38
rs960102817 Chr.22: 38126310-38126310 on Build GRCh38
rs147043645 Chr.22: 38126426-38126426 on Build GRCh38
rs371526199 Chr.22: 38126359-38126359 on Build GRCh38
rs764380622 Chr.22: 38126354-38126354 on Build GRCh38
rs977556550 Chr.22: 38126306-38126306 on Build GRCh38
rs778890839 Chr.22: 38126389-38126389 on Build GRCh38
rs575860043 Chr.22: 38126329-38126329 on Build GRCh38
rs775557315 Chr.22: 38126396-38126396 on Build GRCh38
rs376801641 Chr.22: 38126407-38126407 on Build GRCh38
rs200201397 Chr.22: 38126337-38126337 on Build GRCh38
rs981964953 Chr.22: 38126355-38126355 on Build GRCh38
rs549885315 Chr.22: 38126297-38126297 on Build GRCh38
rs139184008 Chr.22: 38126374-38126374 on Build GRCh38
rs541596845 Chr.22: 38126364-38126364 on Build GRCh38
rs797045888 Chr.22: 38126451-38126451 on Build GRCh38
rs776879486 Chr.22: 38126465-38126465 on Build GRCh38
rs772103372 Chr.22: 38126393-38126393 on Build GRCh38
rs745774208 Chr.22: 38126435-38126435 on Build GRCh38
rs531197938 Chr.22: 38126505-38126505 on Build GRCh38
rs533352113 Chr.22: 38126441-38126441 on Build GRCh38
rs752260146 Chr.22: 38126420-38126420 on Build GRCh38
rs780968037 Chr.22: 38126324-38126324 on Build GRCh38
rs535507569 Chr.22: 38126303-38126303 on Build GRCh38
rs777645662 Chr.22: 38126381-38126381 on Build GRCh38
rs148636646 Chr.22: 38126484-38126484 on Build GRCh38
rs765560166 Chr.22: 38126409-38126409 on Build GRCh38
rs749179801 Chr.22: 38126335-38126335 on Build GRCh38
rs760817038 Chr.22: 38126398-38126398 on Build GRCh38
rs762223777 Chr.22: 38126462-38126462 on Build GRCh38
rs587784329 Chr.22: 38126447-38126447 on Build GRCh38
rs751874222 Chr.22: 38126454-38126463 on Build GRCh38
rs568081052 Chr.22: 38126302-38126302 on Build GRCh38
rs76718524 Chr.22: 38126417-38126417 on Build GRCh38
rs772095529 Chr.22: 38126341-38126341 on Build GRCh38
rs201278654 Chr.22: 38126467-38126467 on Build GRCh38
rs763827059 Chr.22: 38126422-38126422 on Build GRCh38
rs758332232 Chr.22: 38126438-38126438 on Build GRCh38
rs11570708 Chr.22: 38126298-38126298 on Build GRCh38
rs768858806 Chr.22: 38126400-38126400 on Build GRCh38
rs539389084 Chr.22: 38126315-38126315 on Build GRCh38
rs387906864 Chr.22: 38126444-38126444 on Build GRCh38
rs755845373 Chr.22: 38126375-38126375 on Build GRCh38
rs571667445 Chr.22: 38126307-38126307 on Build GRCh38
rs374686605 Chr.22: 38126330-38126330 on Build GRCh38
rs992074994 Chr.22: 38126312-38126312 on Build GRCh38
rs780880188 Chr.22: 38126372-38126372 on Build GRCh38
rs199714835 Chr.22: 38126382-38126382 on Build GRCh38
rs984595225 Chr.22: 38126320-38126320 on Build GRCh38
rs146684391 Chr.22: 38126371-38126371 on Build GRCh38
rs753484966 Chr.22: 38126423-38126423 on Build GRCh38
rs138139283 Chr.22: 38126427-38126427 on Build GRCh38
rs553492349 Chr.22: 38126305-38126305 on Build GRCh38
rs770146189 Chr.22: 38126476-38126476 on Build GRCh38
rs750939090 Chr.22: 38126370-38126370 on Build GRCh38
rs541466301 Chr.22: 38126416-38126416 on Build GRCh38
rs776894280 Chr.22: 38126406-38126406 on Build GRCh38
rs1012889381 Chr.22: 38126440-38126440 on Build GRCh38
rs375874092 Chr.22: 38126384-38126384 on Build GRCh38
rs901501034 Chr.22: 38126342-38126342 on Build GRCh38
rs760921613 Chr.22: 38126353-38126353 on Build GRCh38
rs747074503 Chr.22: 38126445-38126445 on Build GRCh38
rs201801144 Chr.22: 38126390-38126390 on Build GRCh38
rs763451462 Chr.22: 38126490-38126490 on Build GRCh38

View Full Product Details