Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132659307-132659580 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL13
Entrez Gene ID
3596
Gene Name
interleukin 13
Gene Aliases
IL-13, P600
UniGene
Hs.845
Gene Chromosome Location
Chr.5: 132658173-132661109 on Build GRCh38
Transcripts
NM_002188.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6307070 c.206G>T p.S69I Chr.5: 132659449-132659449 on Build GRCh38
3850068 c.224G>A p.G75D Chr.5: 132659467-132659467 on Build GRCh38
6202898 c.179C>T p.P60L Chr.5: 132659422-132659422 on Build GRCh38
109445 c.176C>T p.A59V Chr.5: 132659419-132659419 on Build GRCh38
6684867 c.194G>A p.S65N Chr.5: 132659437-132659437 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143660447 Chr.5: 132659445-132659445 on Build GRCh38
rs200120288 Chr.5: 132659463-132659463 on Build GRCh38
rs1013002320 Chr.5: 132659531-132659531 on Build GRCh38
rs757019938 Chr.5: 132659367-132659367 on Build GRCh38
rs754310137 Chr.5: 132659394-132659397 on Build GRCh38
rs916567856 Chr.5: 132659328-132659328 on Build GRCh38
rs762983915 Chr.5: 132659457-132659457 on Build GRCh38
rs199869056 Chr.5: 132659433-132659433 on Build GRCh38
rs56097948 Chr.5: 132659355-132659355 on Build GRCh38
rs140369292 Chr.5: 132659423-132659423 on Build GRCh38
rs781226872 Chr.5: 132659373-132659373 on Build GRCh38
rs200155765 Chr.5: 132659505-132659505 on Build GRCh38
rs2069746 Chr.5: 132659387-132659387 on Build GRCh38
rs763956913 Chr.5: 132659491-132659491 on Build GRCh38
rs765719901 Chr.5: 132659455-132659455 on Build GRCh38
rs1035481254 Chr.5: 132659521-132659521 on Build GRCh38
rs371914367 Chr.5: 132659426-132659426 on Build GRCh38
rs757139512 Chr.5: 132659487-132659487 on Build GRCh38
rs146770163 Chr.5: 132659422-132659422 on Build GRCh38
rs769401124 Chr.5: 132659377-132659377 on Build GRCh38
rs565434733 Chr.5: 132659329-132659329 on Build GRCh38
rs201923060 Chr.5: 132659508-132659508 on Build GRCh38
rs575619943 Chr.5: 132659346-132659346 on Build GRCh38
rs751745832 Chr.5: 132659466-132659466 on Build GRCh38
rs546150168 Chr.5: 132659479-132659479 on Build GRCh38
rs745785960 Chr.5: 132659376-132659376 on Build GRCh38
rs1025485578 Chr.5: 132659489-132659489 on Build GRCh38
rs200728211 Chr.5: 132659389-132659389 on Build GRCh38
rs148077750 Chr.5: 132659458-132659458 on Build GRCh38
rs200375404 Chr.5: 132659515-132659515 on Build GRCh38
rs748739561 Chr.5: 132659398-132659398 on Build GRCh38
rs1057286474 Chr.5: 132659500-132659500 on Build GRCh38
rs1049129342 Chr.5: 132659439-132659439 on Build GRCh38
rs138680496 Chr.5: 132659419-132659419 on Build GRCh38

View Full Product Details